4-Iodo-5-(trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

059476-250MG
101918-230EA 638.46 USD
101918-230
4-Iodo-5-(trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
4-Iodo-5-(trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₇H₂₄F₃IN₂Si
MDL Number: MFCD18374143
CAS Number: 1261365-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR