5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde

Supplier: Matrix Scientific

059473-250MG
101918-224EA 638.46 USD
101918-224
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde

Hazard: Irritant

Formula: C₉H₅F₃N₂O
MDL Number: MFCD18374140
CAS Number: 1261365-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR