5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-methoxypyridine-3-boronic acid pinacol ester

059452-250MG
101918-182EA 643.08 USD
101918-182
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇BClNO₃
MW: 269.54 g/mol
MDL Number: MFCD09878898
CAS Number: 1083168-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR