5-Chloro-2-methoxypyridin-3-ol

Supplier: Matrix Scientific

059450-250MG
101918-178EA 643.08 USD
101918-178
5-Chloro-2-methoxypyridin-3-ol
5-Chloro-2-methoxypyridin-3-ol

Hazard: Irritant

Formula: C₆H₆ClNO₂
MDL Number: MFCD18374102
CAS Number: 1261365-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR