2,3-Diiodo-5-(trifluoromethyl)pyridine

Supplier: Matrix Scientific

059442-250MG
101918-162EA 643.08 USD
101918-162
2,3-Diiodo-5-(trifluoromethyl)pyridine
2,3-Diiodo-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂F₃I₂N
MDL Number: MFCD16610880
CAS Number: 1227599-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR