2-(Dimethoxymethyl)-6-(trifluoromethyl)-1,8-naphthyridine

Supplier: Matrix Scientific

059441-250MG
101918-160EA 643.08 USD
101918-160
2-(Dimethoxymethyl)-6-(trifluoromethyl)-1,8-naphthyridine
2-(Dimethoxymethyl)-6-(trifluoromethyl)-1,8-naphthyridine

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₂O₂
MDL Number: MFCD18374124
CAS Number: 1261365-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR