3-(Dimethoxymethyl)-1,5-naphthyridine
Supplier: Matrix Scientific
059439-250MG
101918-156EA
643.08
USD
101918-156
3-(Dimethoxymethyl)-1,5-naphthyridine
3-(Dimethoxymethyl)-1,5-naphthyridine
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₂ |
MDL Number:
MFCD18374122 CAS Number: 1261365-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...