(E)-1,5-Naphthyridine-3-carbaldehyde oxime

Supplier: Matrix Scientific

059438-250MG
101918-154EA 643.08 USD
101918-154
(E)-1,5-Naphthyridine-3-carbaldehyde oxime
(E)-1,5-Naphthyridine-3-carbaldehyde oxime

Hazard: Irritant

Formula: C₉H₇N₃O
MDL Number: MFCD18374121
CAS Number: 1261399-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR