5-Chloro-1-(triisopropylsilyl)-4-((trimethylsilyl)ethynyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

059429-250MG
101918-136EA 638.46 USD
101918-136
5-Chloro-1-(triisopropylsilyl)-4-((trimethylsilyl)ethynyl)-1H-pyrrolo[2,3-b]pyridine
5-Chloro-1-(triisopropylsilyl)-4-((trimethylsilyl)ethynyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₂₁H₃₃ClN₂Si₂
MW: 405.14 g/mol
MDL Number: MFCD18374133
CAS Number: 1261365-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR