4-Iodo-1,5-naphthyridin-3-amine

Supplier: Matrix Scientific

059422-250MG
101918-120EA 638.46 USD
101918-120
4-Iodo-1,5-naphthyridin-3-amine
4-Iodo-1,5-naphthyridin-3-amine

Hazard: Irritant

Formula: C₈H₆IN₃
MDL Number: MFCD18374118
CAS Number: 1261365-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR