6-Bromo-7-methoxyfuro[3,2-c]pyridine

Supplier: Matrix Scientific

059421-250MG
101918-118EA 638.46 USD
101918-118
6-Bromo-7-methoxyfuro[3,2-c]pyridine
6-Bromo-7-methoxyfuro[3,2-c]pyridine

Hazard: Irritant

Formula: C₈H₆BrNO₂
MDL Number: MFCD18374152
CAS Number: 1261366-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR