5-Chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylic acid

Supplier: Matrix Scientific

059408-250MG
101918-092EA 553.85 USD
101918-092
5-Chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylic acid
5-Chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD17171345
CAS Number: 1246088-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR