5-Chloro-6-iodo-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid

Supplier: Matrix Scientific

059407-250MG
101918-090EA 638.46 USD
101918-090
5-Chloro-6-iodo-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
5-Chloro-6-iodo-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₄ClIN₂O₂
MDL Number: MFCD17171344
CAS Number: 1246088-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR