2-Bromo-3-methoxypyridin-4-ol

Supplier: Matrix Scientific

059402-250MG
101918-080EA 643.08 USD
101918-080
2-Bromo-3-methoxypyridin-4-ol
2-Bromo-3-methoxypyridin-4-ol

Hazard: Irritant

Formula: C₆H₆BrNO₂
MDL Number: MFCD11553123
CAS Number: 1246088-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR