1,5-Naphthyridine-3-carbaldehyde

Supplier: Matrix Scientific

059395-250MG
101918-066EA 638.46 USD
101918-066
1,5-Naphthyridine-3-carbaldehyde
1,5-Naphthyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₆N₂O
MDL Number: MFCD17171334
CAS Number: 959617-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR