3-Fluoro-5-(3-hydroxyprop-1-yn-1-yl)picolinonitrile

Supplier: Matrix Scientific

059390-250MG
101918-056EA 643.08 USD
101918-056
3-Fluoro-5-(3-hydroxyprop-1-yn-1-yl)picolinonitrile
3-Fluoro-5-(3-hydroxyprop-1-yn-1-yl)picolinonitrile

Hazard: Irritant

Formula: C₉H₅FN₂O
MW: 176.15 g/mol
MDL Number: MFCD17171329
CAS Number: 1246088-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR