2-Chloro-3-iodo-5-(trifluoromethyl)pyridine

Supplier: Matrix Scientific

059388-250MG
101918-052EA 638.46 USD
101918-052
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂ClF₃IN
MDL Number: MFCD08741352
CAS Number: 505084-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR