5-Fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde

Supplier: Matrix Scientific

059386-250MG
101918-048EA 643.08 USD
101918-048
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde

Hazard: Irritant

Formula: C₈H₅FN₂O
MDL Number: MFCD17171327
CAS Number: 1246088-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR