5-Bromo-2-ethyl-3-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

059384-250MG
101918-044EA 638.46 USD
101918-044
5-Bromo-2-ethyl-3-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine
5-Bromo-2-ethyl-3-iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₅H₁₂BrIN₂O₂S
MDL Number: MFCD17171325
CAS Number: 1246088-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR