5-Bromo-2-ethyl-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
Supplier: Matrix Scientific
059378-250MG
101918-032EA
643.08
USD
101918-032
5-Bromo-2-ethyl-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
5-Bromo-2-ethyl-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₁₈H₂₉BrN₂Si |
MDL Number:
MFCD17171321 CAS Number: 1246088-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...