Methyl-5-bromo-3-fluoropicolinate

Synonyms: Methyl 5-bromo-3-fluoropyridine-2-carboxylate

059375-250MG
101918-026EA 261.13 USD
101918-026
Methyl-5-bromo-3-fluoropicolinate
Methyl-5-bromo-3-fluoropicolinate

Hazard: Irritant

Formula: C₇H₅BrFNO₂
MW: 234.03 g/mol
MDL Number: MFCD14698121
CAS Number: 1211538-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR