N-(5-Bromo-2-cyanopyridin-3-yl)pivalamide

Supplier: Matrix Scientific

059374-250MG
101918-024EA 643.08 USD
101918-024
N-(5-Bromo-2-cyanopyridin-3-yl)pivalamide
N-(5-Bromo-2-cyanopyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₂BrN₃O
MDL Number: MFCD17171320
CAS Number: 1246088-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR