7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

059370-250MG
101918-016EA 643.08 USD
101918-016
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD18374089
CAS Number: 1261365-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR