(E)-Methyl-3-(2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)acrylate

Supplier: Matrix Scientific

059369-250MG
101918-014EA 638.46 USD
101918-014
(E)-Methyl-3-(2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)acrylate
(E)-Methyl-3-(2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)acrylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MDL Number: MFCD18374088
CAS Number: 1261399-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR