tert-Butyl (2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)carbamate

Supplier: Matrix Scientific

059368-250MG
101918-012EA 638.46 USD
101918-012
tert-Butyl (2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)carbamate
tert-Butyl (2,3-dihydro[1,4]dioxino[2,3-b]pyridin-7-yl)carbamate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD18374087
CAS Number: 1261365-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR