2,5-Dichloro-6-(hydroxymethyl)-4-iodopyridin-3-ol

Supplier: Matrix Scientific

059366-250MG
101918-008EA 643.08 USD
101918-008
2,5-Dichloro-6-(hydroxymethyl)-4-iodopyridin-3-ol
2,5-Dichloro-6-(hydroxymethyl)-4-iodopyridin-3-ol

Hazard: Irritant

Formula: C₆H₄Cl₂INO₂
MDL Number: MFCD18374112
CAS Number: 1261365-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR