2,5-Dichloro-3-methoxypyridine

059364-250MG
101918-004EA 449.34 USD
101918-004
2,5-Dichloro-3-methoxypyridine
2,5-Dichloro-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MDL Number: MFCD13185540
CAS Number: 1214366-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR