(2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-7-yl)methanol

059358-250MG
101917-992EA 449.34 USD
101917-992
(2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-7-yl)methanol
(2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-7-yl)methanol

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD18261890
CAS Number: 443956-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR