2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-8-ol

Supplier: Matrix Scientific

059352-250MG
101917-980EA 638.46 USD
101917-980
2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-8-ol
2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-8-ol

Hazard: Irritant

Formula: C₇H₇NO₃
MDL Number: MFCD17171314
CAS Number: 1246088-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR