6-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

059351-250MG
101917-978EA 636.92 USD
101917-978
6-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
6-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₆INO₂
MW: 263.04 g/mol
MDL Number: MFCD17171308
CAS Number: 1246088-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR