5-Iodo-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine

Supplier: Matrix Scientific

059347-1G
101917-970EA 913.85 USD
101917-970
5-Iodo-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
5-Iodo-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₁₅H₂₄IN₃Si
MW: 401.37 g/mol
MDL Number: MFCD18374092
CAS Number: 1196662-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR