N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide

059346-250MG
101917-968EA 477.25 USD
101917-968
N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide
N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₂F₃IN₂O
MDL Number: MFCD17171313
CAS Number: 1002916-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR