(E)-2,3-Dihydro-[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime

Supplier: Matrix Scientific

059342-250MG
101917-960EA 638.46 USD
101917-960
(E)-2,3-Dihydro-[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime
(E)-2,3-Dihydro-[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime

Hazard: Irritant

Formula: C₈H₈N₂O₃
MDL Number: MFCD17171311
CAS Number: 1246090-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR