3-Chloro-2-fluoro-4-iodopyridine ≥97%

Supplier: Matrix Scientific

059332-1G
101917-938EA 212.31 USD
101917-938
3-Chloro-2-fluoro-4-iodopyridine ≥97%
3-Chloro-2-fluoro-4-iodopyridine

Hazard: Irritant

Formula: C₅H₂ClFIN
MDL Number: MFCD16610366
CAS Number: 796851-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR