Ethyl-1-[(4-methoxyphenyl)methyl]-4-oxo-1,4-dihydroquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

059306-1G 059306-5G
101917-838EA 1023.08 USD
101917-838 101917-840
Ethyl-1-[(4-methoxyphenyl)methyl]-4-oxo-1,4-dihydroquinoline-3-carboxylate ≥95%
Ethyl-1-[(4-methoxyphenyl)methyl]-4-oxo-1,4-dihydroquinoline-3-carboxylate

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD18426523
CAS Number: 937268-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR