3-Thiophenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

059255-1G
101917-678EA 380 USD
101917-678
3-Thiophenesulfonyl chloride ≥95%
3-Thiophenesulphonyl chloride

Hazard: Irritant

Formula: C₄H₃ClO₂S₂
MDL Number: MFCD02682011
CAS Number: 51175-71-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR