4-Fluorobenzamidoxime ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: (Z)-4-Fluoro-N'-hydroxybenzimidamide

059065-25G 059065-5G
101916-934EA 343.85 USD
101916-934 101916-932
4-Fluorobenzamidoxime ≥95%
4-Fluorobenzamidoxime
Formula: C₇H₇FN₂O
MW: 154.14 g/mol
MDL Number: MFCD00216571
CAS Number: 22179-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR