2-Chlorobenzylideneacetone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Chlorophenyl)but-3-en-2-one

059013-5G 059013-1G
101916-696EA 235.38 USD
101916-696 101916-694
2-Chlorobenzylideneacetone ≥95%
2-Chlorobenzylideneacetone

Hazard: Irritant

Formula: C₁₀H₉ClO
CAS Number: 20766-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR