Cyclopropanecarboxamidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopropanecarboximidamide Hydrochloride

058954-25G 058954-5G
101916-470EA 83.08 USD
101916-470 101916-468
Cyclopropanecarboxamidine hydrochloride ≥95%
Cyclopropanecarboxamidine hydrochloride

Purity: 95+%
CAS: 57297-29-7
MDL Number: MFCD00053010
Molecular Formula: C4H9ClN2
Molecular Weight: 120,58
Hazard: Irritant/Hygroscopic

Formula: C₄H₈N₂
MW: 120.58 g/mol
Melting Pt: 122…123 °C
MDL Number: MFCD00053010
CAS Number: 57297-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR