3-Chlorobenzylideneacetone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Chlorophenyl)but-3-en-2-one

058887-5G
101916-200EA 196.65 USD
101916-200
3-Chlorobenzylideneacetone ≥95%
3-Chlorobenzylideneacetone

Hazard: Irritant

Formula: C₁₀H₉ClO
MW: 180.64
MDL Number: MFCD00052851
CAS Number: 20766-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR