5-Bromo-2,1,3-benzothiadiazole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromobenzo[c][1,2,5]thiadiazole
058770-5G
058770-1G
101915-754EA
197.8
USD
101915-754
101915-752
5-Bromo-2,1,3-benzothiadiazole ≥95%
5-Bromo-2,1,3-benzothiadiazole
Hazard: Irritant
Formula:
C₆H₃BrN₂S MW: 215.0700 g/mol |
MDL Number:
MFCD00460091 CAS Number: 1753-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...