Ethyl-3-(((benzyloxy)carbonyl)(2-ethoxy-2-oxoethyl)amino)propanoate ≥95%

Supplier: Matrix Scientific

058763-1G
101915-716EA 727.69 USD
101915-716
Ethyl-3-(((benzyloxy)carbonyl)(2-ethoxy-2-oxoethyl)amino)propanoate ≥95%
Ethyl-3-(((benzyloxy)carbonyl)(2-ethoxy-2-oxoethyl)amino)propanoate

Purity: 95+%
CAS: 51814-17-6
MDL Number: MFCD16620779
Molecular Formula: C17H23NO6
Molecular Weight: 337,38
Hazard: Irritant

Formula: C₁₇H₂₃NO₆
MDL Number: MFCD16620779
CAS Number: 51814-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR