N-Benzylguanidinium acetate ≥95%

Supplier: Matrix Scientific

058759-1G 058759-5G
101915-704EA 467.69 USD
101915-704 101915-702
N-Benzylguanidinium acetate ≥95%
N-Benzylguanidinium acetate

Purity: 95+%
CAS: 2211-57-6
MDL Number: MFCD09864972
Molecular Formula: C10H15N3O2
Molecular Weight: 209,25
Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MW: 209.25 g/mol
MDL Number: MFCD09864972
CAS Number: 2211-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR