Benzothiazol-4-ylamine, Technical Grade
Supplier: Matrix Scientific
Synonyms:
4-Aminobenzothiazole
, Benzo[d]thiazol-4-amine
058755-1G
058755-500MG
101915-688EA
584.62
USD
101915-688
101915-686
Benzothiazol-4-ylamine, Technical Grade
Benzothiazol-4-ylamine
CAS: 1123-51-9
MDL Number: MFCD00672135
Molecular Formula: C7H6N2S
Molecular Weight: 150,2
Hazard: Irritant/Store in Freezer
Formula:
C₇H₆N₂S MW: 150.2 g/mol |
MDL Number:
MFCD00672135 CAS Number: 1123-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...