Benzo[1,3]dioxole-5-carboxamidoxime ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Hydroxybenzo[d][1,3]dioxole-5-carboximidamide
058753-1G
058753-5G
101915-680EA
204.62
USD
101915-680
101915-682
Benzo[1,3]dioxole-5-carboxamidoxime ≥95%
Benzo[1,3]dioxole-5-carboxamidoxime
Purity: 95+%
CAS: 4720-72-3
MDL Number: MFCD00119015
Molecular Formula: C8H8N2O3
Molecular Weight: 180,16
Hazard: Irritant
Formula:
C₈H₈N₂O₃ MW: 180.16 g/mol |
MDL Number:
MFCD00119015 CAS Number: 4720-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...