1-Benzenesulfonyl-4-chloro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-nitro-1-(phenylsulphonyl)benzene

058751-1G
101915-672EA 281.54 USD
101915-672
1-Benzenesulfonyl-4-chloro-2-nitrobenzene ≥95%
1-Benzenesulfonyl-4-chloro-2-nitrobenzene

Purity: 95+%
CAS: 86030-08-2
MDL Number: MFCD00052250
Molecular Formula: C12H8ClNO4S
Molecular Weight: 297,72
Hazard: Irritant

Formula: C₁₂H₈ClNO₄S
MW: 297.72 g/mol
MDL Number: MFCD00052250
CAS Number: 86030-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR