N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058739-500MG
058739-1G
101915-628EA
284.62
USD
101915-628
101915-630
N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Purity: 95+%
CAS: 159016-25-8
MDL Number: MFCD10568253
Molecular Formula: C12H17N3O2
Molecular Weight: 235,29
Hazard: Irritant
Formula:
C₁₂H₁₇N₃O₂ MW: 235.29 g/mol |
MDL Number:
MFCD10568253 CAS Number: 159016-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...