N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058734-1G
058734-500MG
101915-612EA
376.92
USD
101915-612
101915-610
N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-naphthalen-2-ylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Purity: 95+%
CAS: 1053655-99-4
MDL Number: MFCD10568193
Molecular Formula: C16H19N3O2
Molecular Weight: 285,35
Hazard: Irritant
Formula:
C₁₆H₁₉N₃O₂ MW: 285.35 g/mol |
MDL Number:
MFCD10568193 CAS Number: 1053655-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...