2-Amino-4-methylthiazole-5-carboxylic acid ethylester hydrochloride ≥95%
Supplier: Matrix Scientific
058731-5G
058731-1G
101915-600EA
495.38
USD
101915-600
101915-602
2-Amino-4-methylthiazole-5-carboxylic acid ethylester hydrochloride ≥95%
2-Amino-4-methylthiazole-5-carboxylic acid ethylester hydrochloride
Purity: 95+%
CAS: 53266-92-5
MDL Number: MFCD00829075
Molecular Formula: C7H11ClN2O2S
Molecular Weight: 222,69
Hazard: Irritant
Formula:
C₇H₁₁ClN₂O₂S |
MDL Number:
MFCD00829075 CAS Number: 53266-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...