3-Amino-5-methylpyridine-2-carboxylic acid amide ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-methylpyridine-2-carboxamide
058729-1G
058729-5G
101915-592EA
495.38
USD
101915-592
101915-594
3-Amino-5-methylpyridine-2-carboxylic acid amide ≥95%
3-Amino-5-methylpyridine-2-carboxylic acid amide
Purity: 95+%
CAS: 1208081-47-3
MDL Number: MFCD15142820
Molecular Formula: C7H9N3O
Molecular Weight: 151,17
Hazard: Irritant
Formula:
C₇H₉N₃O MW: 151.17 g/mol |
MDL Number:
MFCD15142820 CAS Number: 1208081-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...